Anti-TYRO3 Mouse Monoclonal Antibody [clone: 6D6F10]

Supplier: Abnova
MAB10417
10743-544EA 428.44 USD
10743-544
Anti-TYRO3 Mouse Monoclonal Antibody [clone: 6D6F10]
Antibodies
Mouse monoclonal antibody raised against partial recombinant TYRO3.

Recommended Dilutions: ELISA: 1:10000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: TYRO3
Clonality: Monoclonal
Clone: 6D6F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR