Anti-VIM Mouse Monoclonal Antibody [clone: 9E7E7,5G3F10,5G3F10E5]

Supplier: Abnova
MAB10418
10744-200EA 428.44 USD
10744-200
Anti-VIM Mouse Monoclonal Antibody [clone: 9E7E7,5G3F10,5G3F10E5]
Antibodies
Mouse monoclonal antibody raised against recombinant VIM.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 9E7E7,5G3F10,5G3F10E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR