Anti-MMP9 Mouse Monoclonal Antibody [clone: 4A3]

Supplier: Abnova
MAB1805
10676-858EA 428 USD
10676-858
Anti-MMP9 Mouse Monoclonal Antibody [clone: 4A3]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of MMP9.

Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR