Anti-PURH Mouse Monoclonal Antibody [clone: F38 P7 H9]

Supplier: Abnova
MAB1824
10673-200EA 428 USD
10673-200
Anti-PURH Mouse Monoclonal Antibody [clone: F38 P7 H9]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of ATIC.

Type: Primary
Antigen: PURH
Clonality: Monoclonal
Clone: F38 P7 H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Frog, Drosophila, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR