Anti-His-Tag Monoclonal Antibody [clone: IPA2C5.1]

Supplier: Abnova
MAB1826
10717-396EA 428 USD
10717-396
Anti-His-Tag Monoclonal Antibody [clone: IPA2C5.1]
Antibodies
Mouse monoclonal antibody raised against His-Tag.

Type: Primary
Antigen: His-Tag
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: IPA2C5.1
Epitope:
Host: IgG2a
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR