Anti-CAPN1 Mouse Monoclonal Antibody [clone: 15C10]

Supplier: Abnova
MAB1837
10704-870EA 428 USD
10704-870
Anti-CAPN1 Mouse Monoclonal Antibody [clone: 15C10]
Antibodies
Mouse monoclonal antibody raised against native CAPN1.

Recommended Dilutions: Western Blot: 0.5-1.0 ?g/ml; ELISA: 0.5-1.0 ?g/ml; Immunoprecipitation: 1-2 ?g/ml

Type: Primary
Antigen: CAPN1
Clonality: Monoclonal
Clone: 15C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR