Anti-IL2 Mouse Monoclonal Antibody [clone: 8D5]

Supplier: Abnova
MAB10421
10719-126EA 345.2 USD
10719-126
Anti-IL2 Mouse Monoclonal Antibody [clone: 8D5]
Antibodies
Mouse monoclonal antibody raised against recombinant IL2.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: 8D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR