Anti-MUSK Mouse Monoclonal Antibody [clone: 10A4]

Supplier: Abnova
MAB10428
10732-862EA 428.44 USD
10732-862
Anti-MUSK Mouse Monoclonal Antibody [clone: 10A4]
Antibodies
Mouse monoclonal antibody raised against recombinant MUSK.

Recommended Dilutions: Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; ELISA: 1:10000

Type: Primary
Antigen: MUSK
Clonality: Monoclonal
Clone: 10A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR