Anti-BDH1 Mouse Monoclonal Antibody [clone: 1A5]

Supplier: Abnova
MAB10451
10691-068EA 428.44 USD
10691-068
Anti-BDH1 Mouse Monoclonal Antibody [clone: 1A5]
Antibodies
Mouse monoclonal antibody raised against recombinant BDH1.

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; ELISA: 1:10000

Type: Primary
Antigen: BDH1
Clonality: Monoclonal
Clone: 1A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR