Anti-BLK Mouse Monoclonal Antibody [clone: 9D10D1]

Supplier: Abnova
MAB10452
10691-338EA 428 USD
10691-338
Anti-BLK Mouse Monoclonal Antibody [clone: 9D10D1]
Antibodies
Mouse monoclonal antibody raised against recombinant BLK.

Recommended Dilutions: Immunohistochemistry: 1:200-1:1000; ELISA: 1:10000

Type: Primary
Antigen: BLK
Clonality: Monoclonal
Clone: 9D10D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR