Anti-CASP3 Mouse Monoclonal Antibody [clone: AM1-4]

Supplier: Abnova
MAB1841
10705-146EA 428 USD
10705-146
Anti-CASP3 Mouse Monoclonal Antibody [clone: AM1-4]
Antibodies
Mouse monoclonal antibody raised against recombinant CASP3.

Type: Primary
Antigen: CASP3
Clonality: Monoclonal
Clone: AM1-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR