Anti-CCND2 Mouse Monoclonal Antibody [clone: DCS3.1]

Supplier: Abnova
MAB1852
10705-762EA 428 USD
10705-762
Anti-CCND2 Mouse Monoclonal Antibody [clone: DCS3.1]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CCND2.

Type: Primary
Antigen: CCND2
Clonality: Monoclonal
Clone: DCS3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR