Anti-CCNE1 Mouse Monoclonal Antibody [clone: 168]

Supplier: Abnova
MAB1856
10705-772EA 428 USD
10705-772
Anti-CCNE1 Mouse Monoclonal Antibody [clone: 168]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CCNE1.

Type: Primary
Antigen: CCNE1
Clonality: Monoclonal
Clone: 168
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR