Anti-CYP3A5 Mouse Monoclonal Antibody [clone: F18 P3 B6]

Supplier: Abnova
MAB1859
10676-120EA 428 USD
10676-120
Anti-CYP3A5 Mouse Monoclonal Antibody [clone: F18 P3 B6]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of CYP3A5.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-5 ?g/ml

Type: Primary
Antigen: CYP3A5
Clonality: Monoclonal
Clone: F18 P3 B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR