Anti-Adenovirus E1A Mouse Monoclonal Antibody [clone: M73]

Supplier: Abnova
MAB1874
10711-760EA 428 USD
10711-760
Anti-Adenovirus E1A Mouse Monoclonal Antibody [clone: M73]
Antibodies
Mouse monoclonal antibody raised against full length recombinant E1A.

Recommended Dilutions: Immunofluorescence: 2-5 ?g/ml; Immunoprecipitation: 1-2 ?g/ml

Type: Primary
Antigen: [delete]
Clonality: Monoclonal
Clone: M73
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR