Anti-CRYAA Mouse Monoclonal Antibody [clone: c9F2]

Supplier: Abnova
MAB1054
10709-162EA 329 USD
10709-162
Anti-CRYAA Mouse Monoclonal Antibody [clone: c9F2]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CRYAA.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: CRYAA
Clonality: Monoclonal
Clone: c9F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR