Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 1G12]

Supplier: Abnova
MAB1063
10688-230EA 329 USD
10688-230
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 1G12]
Antibodies
Mouse monoclonal antibody raised against partial recombinant ADIPOQ.

Recommended Dilutions: Western Blot: 1:250-1:1000

Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: 1G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR