Anti-GST-tag Monoclonal Antibody [clone: KAB29]

Supplier: Abnova
MAB1910
10716-734EA 428 USD
10716-734
Anti-GST-tag Monoclonal Antibody [clone: KAB29]
Antibodies
Mouse monoclonal antibody raised against partial recombinant GST-tag.

Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: GST-tag
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: KAB29
Epitope:
Host: IgG1
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR