Anti-CDK1 Mouse Monoclonal Antibody [clone: POH-1]

Supplier: Abnova
MAB1921
10707-064EA 746.62 USD
10707-064
Anti-CDK1 Mouse Monoclonal Antibody [clone: POH-1]
Antibodies
Mouse monoclonal antibody raised against recombinant CDC2.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50

Type: Primary
Antigen: CDK1
Clonality: Monoclonal
Clone: POH-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR