Anti-PSPH Mouse Monoclonal Antibody [clone: 3G12]

Supplier: Abnova
MAB1093
10730-462EA 329 USD
10730-462
Anti-PSPH Mouse Monoclonal Antibody [clone: 3G12]
Antibodies
Mouse monoclonal antibody raised against full length recombinant PSPH.

Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: PSPH
Clonality: Monoclonal
Clone: 3G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR