Anti-HSPA1A Mouse Monoclonal Antibody [clone: 4E7]

Supplier: Abnova
MAB1094
10721-116EA 329 USD
10721-116
Anti-HSPA1A Mouse Monoclonal Antibody [clone: 4E7]
Antibodies
Mouse monoclonal antibody raised against full length recombinant HSPA1A.

Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 4E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR