Anti-SNAP25 Mouse Monoclonal Antibody [clone: 4E11]

Supplier: Abnova
MAB1105
10738-352EA 329 USD
10738-352
Anti-SNAP25 Mouse Monoclonal Antibody [clone: 4E11]
Antibodies
Mouse monoclonal antibody raised against full length recombinant SNAP25.

Recommended Dilutions: Western Blot: 1:1000-1:3000

Type: Primary
Antigen: SNAP25
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR