Anti-RAB5A Mouse Monoclonal Antibody [clone: 3A4]

Supplier: Abnova
MAB1109
10730-954EA 329 USD
10730-954
Anti-RAB5A Mouse Monoclonal Antibody [clone: 3A4]
Antibodies
Mouse monoclonal antibody raised against full length recombinant RAB5A.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: RAB5A
Clonality: Monoclonal
Clone: 3A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR