Anti-CIB1 Mouse Monoclonal Antibody [clone: 1D1]

Supplier: Abnova
MAB1113
10710-876EA 329 USD
10710-876
Anti-CIB1 Mouse Monoclonal Antibody [clone: 1D1]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CIB1.

Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: CIB1
Clonality: Monoclonal
Clone: 1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR