Anti-HSPD1 Mouse Monoclonal Antibody [clone: 20000]

Supplier: Abnova
MAB1121
10718-264EA 329 USD
10718-264
Anti-HSPD1 Mouse Monoclonal Antibody [clone: 20000]
Antibodies
Mouse monoclonal antibody raised against full length recombinant HSPD1.

Recommended Dilutions: Western Blot: 1:1000-1:3000

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: 20000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR