Anti-MBP Monoclonal Antibody [clone: 10C12]

Supplier: Abnova
MAB1133
10725-080EA 329 USD
10725-080
Anti-MBP Monoclonal Antibody [clone: 10C12]
Antibodies
Mouse monoclonal antibody raised against full length recombinant MBP (Maltose Binding Protein).

Recommended Dilutions: Western Blot: 1:1000-1:3000

Type: Primary
Antigen: MBP
Clonality: Monoclonal
Clone: 10C12
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR