Anti-IKBKG Mouse Monoclonal Antibody [clone: 55AT986.5.78]

Supplier: Abnova
MAB1141
10678-976EA 329 USD
10678-976
Anti-IKBKG Mouse Monoclonal Antibody [clone: 55AT986.5.78]
Antibodies
Mouse monoclonal antibody raised against full length recombinant IKBKG.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: IKBKG
Clonality: Monoclonal
Clone: 55AT986.5.78
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR