Anti-PRDM2 Mouse Monoclonal Antibody [clone: 33AT1045]

Supplier: Abnova
MAB1151
10679-758EA 329 USD
10679-758
Anti-PRDM2 Mouse Monoclonal Antibody [clone: 33AT1045]
Antibodies
Mouse monoclonal antibody raised against partial recombinant PRDM2.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PRDM2
Clonality: Monoclonal
Clone: 33AT1045
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR