Anti-immunodeficiency virus type 1 Gp120 Mouse Monoclonal Antibody [clone: NYRHIV1gp120]

Supplier: Abnova
MAB1263
10720-604EA 346 USD
10720-604
Anti-immunodeficiency virus type 1 Gp120 Mouse Monoclonal Antibody [clone: NYRHIV1gp120]
Antibodies
Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 gp120.

Type: Primary
Antigen: immunodeficiency virus type 1 Gp120
Clonality: Monoclonal
Clone: NYRHIV1gp120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR