Anti-CSF2RA Mouse Monoclonal Antibody [clone: mxsghk-GMCSF]

Supplier: Abnova
MAB1305
10676-084EA 329 USD
10676-084
Anti-CSF2RA Mouse Monoclonal Antibody [clone: mxsghk-GMCSF]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CSF2RA/B.

Recommended Dilutions: ELISA: 1:10000

Type: Primary
Antigen: CSF2RA
Clonality: Monoclonal
Clone: mxsghk-GMCSF
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR