Anti-IFNG Mouse Monoclonal Antibody [clone: IF-H12]

Supplier: Abnova
MAB1306
10676-576EA 329 USD
10676-576
Anti-IFNG Mouse Monoclonal Antibody [clone: IF-H12]
Antibodies
Mouse monoclonal antibody raised against full length recombinant IFNG.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: IF-H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR