Anti-CCL3 Mouse Monoclonal Antibody [clone: mxsghk-1a]

Supplier: Abnova
MAB1313
10673-334EA 329 USD
10673-334
Anti-CCL3 Mouse Monoclonal Antibody [clone: mxsghk-1a]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CCL3.

Recommended Dilutions: ELISA: 1:20000

Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: mxsghk-1a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR