Anti-I-Ak Mouse Monoclonal Antibody [clone: IA-G3]

Supplier: Abnova
MAB1333
10676-552EA 329 USD
10676-552
Anti-I-Ak Mouse Monoclonal Antibody [clone: IA-G3]
Antibodies
Mouse monoclonal antibody raised against native I-Ak.

Type: Primary
Antigen: I-Ak
Clonality: Monoclonal
Clone: IA-G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR