Anti-I-Ek Mouse Monoclonal Antibody [clone: IE-D6]

Supplier: Abnova
MAB1335
10676-564EA 329 USD
10676-564
Anti-I-Ek Mouse Monoclonal Antibody [clone: IE-D6]
Antibodies
Mouse monoclonal antibody raised against native I-Ek.

Type: Primary
Antigen: I-Ek
Clonality: Monoclonal
Clone: IE-D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR