Anti-Neisseria gonorrhoeae Mouse Monoclonal Antibody [clone: MS-20]

Supplier: Abnova
MAB1337
10681-176EA 406.15 USD
10681-176
Anti-Neisseria gonorrhoeae Mouse Monoclonal Antibody [clone: MS-20]
Antibodies
Mouse monoclonal antibody raised against Neisseria gonorrhoeae.

Type: Primary
Antigen: Neisseria gonorrhoeae
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: MS-20
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR