Anti-immunodeficiency virus type 1 p24 Mouse Monoclonal Antibody [clone: 32]

Supplier: Abnova
MAB1338
10681-944EA 329 USD
10681-944
Anti-immunodeficiency virus type 1 p24 Mouse Monoclonal Antibody [clone: 32]
Antibodies
Mouse monoclonal antibody raised against full length recombinant human immunodeficiency virus type 1 p24.

Type: Primary
Antigen: immunodeficiency virus type 1 p24
Clonality: Monoclonal
Clone: 32
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR