Anti-immunodeficiency virus type 1 p24 Mouse Monoclonal Antibody [clone: 49]

Supplier: Abnova
MAB1342
10681-952EA 329 USD
10681-952
Anti-immunodeficiency virus type 1 p24 Mouse Monoclonal Antibody [clone: 49]
Antibodies
Mouse monoclonal antibody raised against full length recombinant human immunodeficiency virus type 1 p24.

Type: Primary
Antigen: immunodeficiency virus type 1 p24
Clonality: Monoclonal
Clone: 49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR