Anti-immunodeficiency virus type 1 env Mouse Monoclonal Antibody [clone: HG-4]

Supplier: Abnova
MAB1343
10676-194EA 329 USD
10676-194
Anti-immunodeficiency virus type 1 env Mouse Monoclonal Antibody [clone: HG-4]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of human immunodeficiency virus type 1 env.

Type: Primary
Antigen: immunodeficiency virus type 1 env
Clonality: Monoclonal
Clone: HG-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR