Anti-immunodeficiency virus type 1 gp41 Mouse Monoclonal Antibody [clone: YR-12]

Supplier: Abnova
MAB1344
10676-324EA 329 USD
10676-324
Anti-immunodeficiency virus type 1 gp41 Mouse Monoclonal Antibody [clone: YR-12]
Antibodies
Mouse monoclonal antibody raised against full length recombinant human immunodeficiency virus type 1 gp41.

Type: Primary
Antigen: immunodeficiency virus type 1 gp41
Clonality: Monoclonal
Clone: YR-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR