Anti-immunodeficiency virus-2 gp36 Mouse Monoclonal Antibody [clone: YR-6]

Supplier: Abnova
MAB1345
10676-322EA 329 USD
10676-322
Anti-immunodeficiency virus-2 gp36 Mouse Monoclonal Antibody [clone: YR-6]
Antibodies
Mouse monoclonal antibody raised against full length recombinant human immunodeficiency virus type 2 gp36.

Type: Primary
Antigen: immunodeficiency virus-2 gp36
Clonality: Monoclonal
Clone: YR-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR