Anti-AKR1C1 Mouse Monoclonal Antibody [clone: AT6D1]

Supplier: Abnova
MAB10008
10688-650EA 329 USD
10688-650
Anti-AKR1C1 Mouse Monoclonal Antibody [clone: AT6D1]
Antibodies
Mouse monoclonal antibody raised against full length recombinant AKR1C1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: AKR1C1
Clonality: Monoclonal
Clone: AT6D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR