Anti-NRAS Mouse Monoclonal Antibody [clone: AT2G9]

Supplier: Abnova
MAB10009
10727-648EA 329 USD
10727-648
Anti-NRAS Mouse Monoclonal Antibody [clone: AT2G9]
Antibodies
Mouse monoclonal antibody raised against full length recombinant NRAS.

Recommended Dilutions: Western Blot: 1:500-1:5000

Type: Primary
Antigen: NRAS
Clonality: Monoclonal
Clone: AT2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR