Anti-PAX8 Rabbit Monoclonal Antibody [clone: ZR-1]

Supplier: Abnova
MAB10020
10735-544EA 1061 USD
10735-544
Anti-PAX8 Rabbit Monoclonal Antibody [clone: ZR-1]
Antibodies
Rabbit monoclonal antibody raised against synthetic peptide of PAX8.

Recommended Dilutions: Recommend dilution at 1:50-1:100

Type: Primary
Antigen: PAX8
Clonality: Monoclonal
Clone: ZR-1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR