Anti-Thy1 Rat Monoclonal Antibody [clone: T-B8]

Supplier: Abnova
MAB1358
10681-990EA 346 USD
10681-990
Anti-Thy1 Rat Monoclonal Antibody [clone: T-B8]
Antibodies
Rat monoclonal antibody raised against native Thy1.

Type: Primary
Antigen: Thy1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: T-B8
Epitope:
Host: IgG
Isotype: Rat
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR