Anti-MHC1 Mouse Monoclonal Antibody [clone: KD-H3]

Supplier: Abnova
MAB1363
10676-844EA 329 USD
10676-844
Anti-MHC1 Mouse Monoclonal Antibody [clone: KD-H3]
Antibodies
Mouse monoclonal antibody raised against native MHC Class I.

Type: Primary
Antigen: MHC1
Clonality: Monoclonal
Clone: KD-H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR