Anti-AKT1 Mouse Monoclonal Antibody [clone: M11]

Supplier: Abnova
MAB1364
10688-668EA 329 USD
10688-668
Anti-AKT1 Mouse Monoclonal Antibody [clone: M11]
Antibodies
Mouse monoclonal antibody raised against partial recombinant AKT1.

Recommended Dilutions: ELISA: 1:2000; Immunocytochemistry: 1:250; Western Blot: 1:1000

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: M11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR