Anti-CDC42 Mouse Monoclonal Antibody [clone: M152]

Supplier: Abnova
MAB1376
10707-146EA 329 USD
10707-146
Anti-CDC42 Mouse Monoclonal Antibody [clone: M152]
Antibodies
Mouse monoclonal antibody raised against full length recombinant Cdc42.

Recommended Dilutions: ELISA: 1:2000; Immunocytochemistry: 1:50; Western Blot: 1:500

Type: Primary
Antigen: CDC42
Clonality: Monoclonal
Clone: M152
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR