Anti-MAPK14 Mouse Monoclonal Antibody [clone: M138]

Supplier: Abnova
MAB1384
10732-492EA 427.45 USD
10732-492
Anti-MAPK14 Mouse Monoclonal Antibody [clone: M138]
Antibodies
Mouse monoclonal antibody raised against partial recombinant Mapk14.

Recommended Dilutions: ELISA: 1:2000; Immunocytochemistry: 1:500; Western Blot: 1:1000

Type: Primary
Antigen: MAPK14
Clonality: Monoclonal
Clone: M138
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR