Anti-LTF Mouse Monoclonal Antibody [clone: NI 25]

Supplier: Abnova
MAB10070
10676-770EA 530.63 USD
10676-770
Anti-LTF Mouse Monoclonal Antibody [clone: NI 25]
Antibodies
Mouse monoclonal antibody raised against LTF.

Recommended Dilutions: ELISA: 1:50; Western Blot: 1:250; Immunohistochemistry: 1:20

Type: Primary
Antigen: LTF
Clonality: Monoclonal
Clone: NI 25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR