Anti-LTF Mouse Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate)) [clone: NI 25]

Supplier: Abnova
MAB10072
10676-778EA 747 USD
10676-778
Anti-LTF Mouse Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate)) [clone: NI 25]
Antibodies
Mouse monoclonal antibody raised against LTF.

Recommended Dilutions: Immunohistochemistry: 1:10-1:100

Type: Primary
Antigen: LTF
Clonality: Monoclonal
Clone: NI 25
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR